Assay Details
| Gene Symbol: ints-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307283.3 | NP_001294212.1 | 8 - 9 | 1872 | 98 | |
| NM_001307284.2 | NP_001294213.1 | 8 - 9 | 1544 | 98 | ||
More Information
Set Membership: |
|

ints-6
Genomic Map