Assay Details
| Gene Symbol: parg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069095.5 | NP_501496.2 | 7 - 8 | 1334 | 91 | |
| GenBank mRNA | AF548468.1 | - | 7 - 8 | 1294 | 91 | |
More Information
Set Membership: |
|

parg-2
Genomic Map