Assay Details
| Gene Symbol: parg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069095.5 | NP_501496.2 | 8 - 9 | 1458 | 109 | |
| GenBank mRNA | AF548468.1 | - | 8 - 9 | 1418 | 109 | |
More Information
Set Membership: |
|

parg-2
Genomic Map