Assay Details
| Gene Symbol: parg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027964.4 | NP_001023135.1 | 11 - 12 | 1823 | 117 | |
| NM_001027965.4 | NP_001023136.1 | 10 - 11 | 1774 | 117 | ||
| NM_001129300.4 | NP_001122772.1 | 11 - 12 | 1785 | 117 | ||
| NM_001268395.3 | NP_001255324.1 | 9 - 10 | 1254 | 117 | ||
| NM_001276825.3 | NP_001263754.1 | 11 - 12 | 1828 | 117 | ||
| NM_001307131.4 | NP_001294060.1 | 10 - 11 | 1762 | 117 | ||
| NM_001307132.3 | NP_001294061.1 | 11 - 12 | 1819 | 117 | ||
| NM_001307133.3 | NP_001294062.1 | 11 - 12 | 1834 | 117 | ||
| NM_001380380.1 | NP_001367109.1 | 10 - 11 | 1320 | 117 | ||
| GenBank mRNA | AY185493.1 | - | 11 - 12 | 1845 | 117 | |
| AY185494.1 | - | 10 - 11 | 1794 | 117 | ||
More Information
Set Membership: |
|

parg-1
Genomic Map