Assay Details
| Gene Symbol: parg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027964.4 | NP_001023135.1 | 12 - 13 | 2110 | 114 | |
| NM_001027965.4 | NP_001023136.1 | 11 - 12 | 2061 | 114 | ||
| NM_001268395.3 | NP_001255324.1 | 10 - 11 | 1541 | 114 | ||
| NM_001276825.3 | NP_001263754.1 | 12 - 13 | 2115 | 114 | ||
| NM_001307131.4 | NP_001294060.1 | 11 - 12 | 2049 | 114 | ||
| NM_001307132.3 | NP_001294061.1 | 12 - 13 | 2106 | 114 | ||
| NM_001307133.3 | NP_001294062.1 | 12 - 13 | 2121 | 114 | ||
| NM_001380380.1 | NP_001367109.1 | 11 - 12 | 1607 | 114 | ||
| GenBank mRNA | AY185493.1 | - | 12 - 13 | 2132 | 114 | |
| AY185494.1 | - | 11 - 12 | 2081 | 114 | ||
More Information
Set Membership: |
|

parg-1
Genomic Map