Assay Details
| Gene Symbol: parg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027964.4 | NP_001023135.1 | 1 - 2 | 84 | 61 | |
| NM_001027965.4 | NP_001023136.1 | 1 - 2 | 86 | 61 | ||
| NM_001129300.4 | NP_001122772.1 | 1 - 2 | 46 | 61 | ||
| NM_001276825.3 | NP_001263754.1 | 1 - 2 | 86 | 61 | ||
| NM_001307131.4 | NP_001294060.1 | 1 - 2 | 83 | 61 | ||
| NM_001307132.3 | NP_001294061.1 | 1 - 2 | 86 | 61 | ||
| NM_001307133.3 | NP_001294062.1 | 1 - 2 | 86 | 61 | ||
| GenBank mRNA | AY185493.1 | - | 1 - 2 | 106 | 61 | |
| AY185494.1 | - | 1 - 2 | 106 | 61 | ||
More Information
Set Membership: |
|

parg-1
Genomic Map