Assay Details
| Gene Symbol: parg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027964.4 | NP_001023135.1 | 7 - 8 | 1130 | 148 | |
| NM_001027965.4 | NP_001023136.1 | 6 - 7 | 1081 | 148 | ||
| NM_001129300.4 | NP_001122772.1 | 7 - 8 | 1092 | 148 | ||
| NM_001268395.3 | NP_001255324.1 | 5 - 6 | 561 | 148 | ||
| NM_001276825.3 | NP_001263754.1 | 7 - 8 | 1135 | 148 | ||
| NM_001307131.4 | NP_001294060.1 | 6 - 7 | 1069 | 148 | ||
| NM_001307132.3 | NP_001294061.1 | 7 - 8 | 1126 | 148 | ||
| NM_001307133.3 | NP_001294062.1 | 7 - 8 | 1141 | 148 | ||
| NM_001380380.1 | NP_001367109.1 | 6 - 7 | 627 | 148 | ||
| GenBank mRNA | AY185493.1 | - | 7 - 8 | 1152 | 148 | |
| AY185494.1 | - | 6 - 7 | 1101 | 148 | ||
More Information
Set Membership: |
|

parg-1
Genomic Map