Assay Details
| Gene Symbol: alp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028199.5 | NP_001023370.1 | 6 - 7 | 667 | 83 | |
| NM_001028200.6 | NP_001023371.1 | 7 - 8 | 878 | 83 | ||
| NM_001047522.4 | NP_001040987.1 | 7 - 8 | 884 | 83 | ||
| NM_001380387.1 | NP_001367079.1 | 7 - 8 | 878 | 83 | ||
| NM_069133.5 | NP_501534.2 | 7 - 8 | 878 | 83 | ||
More Information
Set Membership: |
|

alp-1
Genomic Map