Assay Details
| Gene Symbol: spr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026161.2 | NP_001021332.1 | 1 - 2 | 525 | 66 | |
| GenBank mRNA | AF321546.1 | - | 1 - 2 | 502 | 66 | |
More Information
Set Membership: |
|

spr-2
Genomic Map