Assay Details
| Gene Symbol: slc-25A26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069151.5 | NP_501552.1 | 1 - 2 | 154 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc-25A26| Gene Symbol: slc-25A26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069151.5 | NP_501552.1 | 1 - 2 | 154 | 70 | |
Set Membership: |
|