Assay Details
| Gene Symbol: bre-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069163.5 | NP_501564.1 | 1 - 2 | 134 | 97 | |
| GenBank mRNA | AM231683.1 | - | 1 - 2 | 69 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bre-1| Gene Symbol: bre-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069163.5 | NP_501564.1 | 1 - 2 | 134 | 97 | |
| GenBank mRNA | AM231683.1 | - | 1 - 2 | 69 | 97 | |
Set Membership: |
|