Assay Details
| Gene Symbol: mltn-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047465.4 | NP_001040930.2 | 9 - 10 | 1802 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mltn-12| Gene Symbol: mltn-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047465.4 | NP_001040930.2 | 9 - 10 | 1802 | 91 | |
Set Membership: |
|