Assay Details
| Gene Symbol: wnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268439.4 | NP_001255368.1 | 11 - 12 | 4785 | 97 | |
| NM_001268441.3 | NP_001255370.1 | 11 - 12 | 4784 | 97 | ||
| NM_001268443.3 | NP_001255372.1 | 10 - 11 | 4458 | 97 | ||
| NM_001268445.3 | NP_001255374.1 | 8 - 9 | 4305 | 97 | ||
| NM_001268446.3 | NP_001255375.1 | 8 - 9 | 4296 | 97 | ||
| NM_001268447.3 | NP_001255376.1 | 8 - 9 | 4299 | 97 | ||
| NM_001268448.3 | NP_001255377.1 | 8 - 9 | 4290 | 97 | ||
| NM_001307103.3 | NP_001294032.1 | 11 - 12 | 4789 | 97 | ||
| NM_001307107.3 | NP_001294036.1 | 11 - 12 | 4779 | 97 | ||
| NM_001380399.1 | NP_001368454.1 | 11 - 12 | 4776 | 97 | ||
| NM_001380400.1 | NP_001368092.1 | 11 - 12 | 4770 | 97 | ||
| NM_069202.6 | NP_501603.3 | 11 - 12 | 4764 | 97 | ||
More Information
Set Membership: |
|

wnk-1
Genomic Map