Assay Details
| Gene Symbol: F49C12.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069226.2 | NP_501627.1 | 6 - 7 | 651 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F49C12.5| Gene Symbol: F49C12.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069226.2 | NP_501627.1 | 6 - 7 | 651 | 145 | |
Set Membership: |
|