Assay Details
| Gene Symbol: coa-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028220.4 | NP_001023391.1 | 3 - 4 | 430 | 110 | |
| NM_001383161.1 | 3 - 4 | 411 | 110 | |||
| Gene Symbol: T20D3.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383162.1 | 4 - 5 | 556 | 110 | ||
More Information
Set Membership: |
|

coa-4
Genomic Map