Assay Details
| Gene Symbol: coa-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028220.4 | NP_001023391.1 | 7 - 8 | 990 | 79 | |
| NM_001383161.1 | 6 - 7 | 872 | 79 | |||
| Gene Symbol: T20D3.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383162.1 | 8 - 9 | 1116 | 79 | ||
More Information
Set Membership: |
|

coa-4
Genomic Map