Assay Details
| Gene Symbol: pigc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069244.3 | NP_501645.1 | 2 - 3 | 537 | 178 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pigc-1| Gene Symbol: pigc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069244.3 | NP_501645.1 | 2 - 3 | 537 | 178 | |
Set Membership: |
|