Assay Details
| Gene Symbol: del-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268482.3 | NP_001255411.1 | 5 - 6 | 511 | 180 | |
| NM_001380416.1 | NP_001367239.1 | 1 - 2 | 93 | 180 | ||
More Information
Set Membership: |
|

del-2
Genomic Map