Assay Details
| Gene Symbol: del-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268482.3 | NP_001255411.1 | 8 - 9 | 1149 | 129 | |
| NM_001380416.1 | NP_001367239.1 | 4 - 5 | 731 | 129 | ||
More Information
Set Membership: |
|

del-2
Genomic Map