Assay Details
| Gene Symbol: acc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306936.3 | NP_001293865.1 | 11 - 12 | 1305 | 107 | |
| NM_069314.7 | NP_501715.1 | 12 - 13 | 1414 | 107 | ||
More Information
Set Membership: |
|

acc-1
Genomic Map