Assay Details
| Gene Symbol: acc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306936.3 | NP_001293865.1 | 1 - 2 | 152 | 118 | |
| NM_069314.7 | NP_501715.1 | 2 - 3 | 261 | 118 | ||
More Information
Set Membership: |
|

acc-1
Genomic Map