Assay Details
| Gene Symbol: fat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268494.2 | NP_001255423.1 | 3 - 4 | 636 | 71 | |
| NM_001268495.3 | NP_001255424.1 | 1 - 2 | 321 | 71 | ||
| GenBank mRNA | AF078796.1 | - | 3 - 4 | 594 | 71 | |
| AF114440.1 | - | 3 - 4 | 636 | 71 | ||
More Information
Set Membership: |
|

fat-4
Genomic Map