Assay Details
| Gene Symbol: fat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268494.2 | NP_001255423.1 | 4 - 5 | 925 | 112 | |
| NM_001268495.3 | NP_001255424.1 | 2 - 3 | 610 | 112 | ||
| GenBank mRNA | AF078796.1 | - | 4 - 5 | 883 | 112 | |
| AF114440.1 | - | 4 - 5 | 925 | 112 | ||
More Information
Set Membership: |
|

fat-4
Genomic Map