Assay Details
| Gene Symbol: fat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268497.2 | NP_001255426.1 | 3 - 4 | 652 | 153 | |
| NM_001307156.3 | NP_001294085.1 | 1 - 2 | 309 | 153 | ||
More Information
Set Membership: |
|

fat-3
Genomic Map