Assay Details
| Gene Symbol: fat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268497.2 | NP_001255426.1 | 6 - 7 | 1192 | 98 | |
| NM_001268498.3 | NP_001255427.1 | 2 - 3 | 378 | 98 | ||
| NM_001307156.3 | NP_001294085.1 | 4 - 5 | 849 | 98 | ||
More Information
Set Membership: |
|

fat-3
Genomic Map