Assay Details
| Gene Symbol: kin-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069360.3 | NP_501761.2 | 2 - 3 | 769 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-24| Gene Symbol: kin-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069360.3 | NP_501761.2 | 2 - 3 | 769 | 89 | |
Set Membership: |
|