Assay Details
| Gene Symbol: gon-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268519.3 | NP_001255448.1 | 22 - 23 | 5291 | 89 | |
| NM_001372913.1 | NP_001359558.1 | 24 - 25 | 5750 | 89 | ||
| NR_056474.1 | 24 - 25 | 5746 | 89 | |||
More Information
Set Membership: |
|

gon-1
Genomic Map