Assay Details
| Gene Symbol: gon-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268519.3 | NP_001255448.1 | 2 - 3 | 151 | 86 | |
| NM_001372913.1 | NP_001359558.1 | 5 - 6 | 655 | 86 | ||
| NR_056474.1 | 5 - 6 | 651 | 86 | |||
More Information
Set Membership: |
|

gon-1
Genomic Map