Assay Details
| Gene Symbol: gon-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268519.3 | NP_001255448.1 | 10 - 11 | 1708 | 90 | |
| NM_001372913.1 | NP_001359558.1 | 13 - 14 | 2212 | 90 | ||
| NR_056474.1 | 13 - 14 | 2208 | 90 | |||
More Information
Set Membership: |
|

gon-1
Genomic Map