Assay Details
| Gene Symbol: tsp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069452.5 | NP_501853.1 | 1 - 2 | 108 | 145 | |
| Gene Symbol: pigs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383182.1 | 1 - 2 | 108 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383182.1 transcripts.
Set Membership: |
|

tsp-12
Genomic Map