Assay Details
| Gene Symbol: ttr-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380433.1 | NP_001366759.1 | 3 - 4 | 913 | 73 | |
| Gene Symbol: copg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047523.5 | NP_001040988.1 | 3 - 4 | 946 | 73 | |
| NM_001047524.5 | NP_001040989.1 | 3 - 4 | 961 | 73 | ||
| NM_001268535.4 | NP_001255464.1 | 3 - 4 | 969 | 73 | ||
| NM_001307032.3 | NP_001293961.1 | 2 - 3 | 843 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380433.1 transcripts.
Set Membership: |
|

ttr-54
Genomic Map