Assay Details
| Gene Symbol: ttr-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380433.1 | NP_001366759.1 | 5 - 6 | 2316 | 66 | |
| Gene Symbol: copg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047523.5 | NP_001040988.1 | 5 - 6 | 2349 | 66 | |
| NM_001047524.5 | NP_001040989.1 | 5 - 6 | 2364 | 66 | ||
| NM_001268535.4 | NP_001255464.1 | 5 - 6 | 2372 | 66 | ||
| NM_001307032.3 | NP_001293961.1 | 4 - 5 | 2246 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380433.1 transcripts.
Set Membership: |
|

ttr-54
Genomic Map