Assay Details
| Gene Symbol: pps-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307031.3 | NP_001293960.1 | 2 - 3 | 272 | 74 | |
| NM_069456.5 | NP_501857.1 | 2 - 3 | 270 | 74 | ||
More Information
Set Membership: |
|

pps-1
Genomic Map