Assay Details
| Gene Symbol: pps-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307031.3 | NP_001293960.1 | 4 - 5 | 1633 | 88 | |
| NM_069456.5 | NP_501857.1 | 4 - 5 | 1625 | 88 | ||
More Information
Set Membership: |
|

pps-1
Genomic Map