Assay Details
| Gene Symbol: ebax-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069487.6 | NP_501888.2 | 8 - 9 | 4602 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ebax-1| Gene Symbol: ebax-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069487.6 | NP_501888.2 | 8 - 9 | 4602 | 98 | |
Set Membership: |
|