Assay Details
| Gene Symbol: iars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380444.2 | NP_001366774.1 | 5 - 6 | 1866 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

iars-1| Gene Symbol: iars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380444.2 | NP_001366774.1 | 5 - 6 | 1866 | 68 | |
Set Membership: |
|