Assay Details
| Gene Symbol: spe-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069518.4 | NP_501919.1 | 5 - 6 | 1578 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spe-26| Gene Symbol: spe-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069518.4 | NP_501919.1 | 5 - 6 | 1578 | 99 | |
Set Membership: |
|