Assay Details
| Gene Symbol: mec-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027940.5 | NP_001023111.1 | 7 - 8 | 999 | 67 | |
| NM_001136380.4 | NP_001129852.1 | 5 - 6 | 769 | 67 | ||
| NM_001356932.4 | NP_001343802.1 | 7 | 944 | 67 | ||
| GenBank mRNA | L02877.1 | - | 7 - 8 | 966 | 67 | |
More Information
Set Membership: |
|

mec-3
Genomic Map