Assay Details
| Gene Symbol: cal-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268562.3 | NP_001255491.1 | 2 - 3 | 438 | 134 | |
| NM_001383190.1 | 6 - 7 | 845 | 134 | |||
| NM_001383191.1 | 5 - 6 | 698 | 134 | |||
More Information
Set Membership: |
|

cal-4
Genomic Map