Assay Details
| Gene Symbol: zim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136397.4 | NP_001129869.1 | 4 - 5 | 1386 | 87 | |
| NM_001276847.3 | NP_001263776.1 | 1 - 2 | 238 | 87 | ||
| NM_001276848.3 | NP_001263777.1 | 1 - 2 | 238 | 87 | ||
| NM_069547.4 | NP_501948.2 | 4 - 5 | 1384 | 87 | ||
More Information
Set Membership: |
|

zim-1
Genomic Map