Assay Details
| Gene Symbol: zim-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307056.3 | NP_001293985.1 | 7 - 8 | 2465 | 85 | |
| NM_069552.3 | NP_501953.2 | 4 - 5 | 1339 | 85 | ||
| Gene Symbol: him-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383193.1 | 4 - 5 | 1336 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383193.1 transcripts.
Set Membership: |
|

zim-3
Genomic Map