Assay Details
| Gene Symbol: nkb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069557.4 | NP_501958.1 | 2 - 3 | 278 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nkb-2| Gene Symbol: nkb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069557.4 | NP_501958.1 | 2 - 3 | 278 | 71 | |
Set Membership: |
|