Assay Details
| Gene Symbol: ptr-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069562.4 | NP_501963.1 | 4 - 5 | 1079 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-14| Gene Symbol: ptr-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069562.4 | NP_501963.1 | 4 - 5 | 1079 | 124 | |
Set Membership: |
|