Assay Details
| Gene Symbol: fmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069571.5 | NP_501972.1 | 2 - 3 | 185 | 97 | |
| GenBank mRNA | AJ582071.1 | - | 2 - 3 | 185 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fmo-2| Gene Symbol: fmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069571.5 | NP_501972.1 | 2 - 3 | 185 | 97 | |
| GenBank mRNA | AJ582071.1 | - | 2 - 3 | 185 | 97 | |
Set Membership: |
|