Assay Details
| Gene Symbol: pyk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268586.2 | NP_001255515.1 | 3 - 4 | 759 | 108 | |
| NM_001268587.3 | NP_001255516.1 | 2 - 3 | 828 | 108 | ||
| NM_001268588.3 | NP_001255517.1 | 2 - 3 | 566 | 108 | ||
More Information
Set Membership: |
|

pyk-2
Genomic Map