Assay Details
| Gene Symbol: pyk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268586.2 | NP_001255515.1 | 6 - 7 | 1415 | 89 | |
| NM_001268587.3 | NP_001255516.1 | 5 - 6 | 1484 | 89 | ||
| NM_001268588.3 | NP_001255517.1 | 5 - 6 | 1222 | 89 | ||
More Information
Set Membership: |
|

pyk-2
Genomic Map