Assay Details
| Gene Symbol: dnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268592.3 | NP_001255521.1 | 11 - 12 | 2747 | 104 | |
| NM_001268593.3 | NP_001255522.1 | 5 - 6 | 1860 | 104 | ||
| NM_001276852.4 | NP_001263781.1 | 9 - 10 | 2545 | 104 | ||
| NM_001276853.3 | NP_001263782.1 | 10 - 11 | 2663 | 104 | ||
| NM_001383200.1 | 11 - 12 | 2792 | 104 | |||
More Information
Set Membership: |
|

dnc-1
Genomic Map