Assay Details
| Gene Symbol: dnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268592.3 | NP_001255521.1 | 2 - 3 | 413 | 61 | |
| NM_001276852.4 | NP_001263781.1 | 2 - 3 | 370 | 61 | ||
| NM_001276853.3 | NP_001263782.1 | 2 - 3 | 413 | 61 | ||
| NM_001383200.1 | 3 - 4 | 533 | 61 | |||
More Information
Set Membership: |
|

dnc-1
Genomic Map