Assay Details
| Gene Symbol: dnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268592.3 | NP_001255521.1 | 7 - 8 | 955 | 73 | |
| NM_001268593.3 | NP_001255522.1 | 1 - 2 | 68 | 73 | ||
| NM_001276852.4 | NP_001263781.1 | 5 - 6 | 753 | 73 | ||
| NM_001276853.3 | NP_001263782.1 | 6 - 7 | 871 | 73 | ||
| NM_001383200.1 | 7 - 8 | 1000 | 73 | |||
More Information
Set Membership: |
|

dnc-1
Genomic Map