Assay Details
| Gene Symbol: dnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268592.3 | NP_001255521.1 | 10 - 11 | 2301 | 69 | |
| NM_001268593.3 | NP_001255522.1 | 4 - 5 | 1414 | 69 | ||
| NM_001276852.4 | NP_001263781.1 | 8 - 9 | 2099 | 69 | ||
| NM_001276853.3 | NP_001263782.1 | 9 - 10 | 2217 | 69 | ||
| NM_001383200.1 | 10 - 11 | 2346 | 69 | |||
More Information
Set Membership: |
|

dnc-1
Genomic Map